siRNA Details
Gene Symbol: Olfr1497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146741.1 | NP_666952.1 | 1 | 138 | ||
GenBank mRNA | BC111435.1 | - | 1 | 136 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146741.1 | NP_666952.1 | 1 | 138 | ||
GenBank mRNA | BC111435.1 | - | 1 | 136 |