siRNA Details
| Gene Symbol: Olfr678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146758.1 | NP_666969.1 | 1 | 283 | ||
| GenBank mRNA | BC128570.1 | - | 1 | 285 | 40135306 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr678| Gene Symbol: Olfr678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146758.1 | NP_666969.1 | 1 | 283 | ||
| GenBank mRNA | BC128570.1 | - | 1 | 285 | 40135306 | |