siRNA Details
| Gene Symbol: Olfr920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146787.2 | NP_666998.2 | 2 | 1388 | ||
| GenBank mRNA | AK040176.1 | - | 1 | 535 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr920| Gene Symbol: Olfr920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146787.2 | NP_666998.2 | 2 | 1388 | ||
| GenBank mRNA | AK040176.1 | - | 1 | 535 | ||