siRNA Details
| Gene Symbol: Olfr716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146604.1 | NP_666815.1 | 1 | 308 | ||
| GenBank mRNA | BC120510.1 | - | 1 | 400 | 40129433 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr716| Gene Symbol: Olfr716 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146604.1 | NP_666815.1 | 1 | 308 | ||
| GenBank mRNA | BC120510.1 | - | 1 | 400 | 40129433 | |