siRNA Details
| Gene Symbol: Olfr418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146651.2 | NP_666862.2 | 1 | 845 | ||
| GenBank mRNA | BC119223.1 | - | 1 | 845 | 8733972 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr418| Gene Symbol: Olfr418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146651.2 | NP_666862.2 | 1 | 845 | ||
| GenBank mRNA | BC119223.1 | - | 1 | 845 | 8733972 | |