siRNA Details
| Gene Symbol: Olfr491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146736.1 | NP_666947.1 | 1 | 913 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr491| Gene Symbol: Olfr491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146736.1 | NP_666947.1 | 1 | 913 | ||