siRNA Details
Gene Symbol: Olfr494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146737.1 | NP_666948.1 | 1 | 317 | ||
GenBank mRNA | BC127987.1 | - | 321 | 40135591 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146737.1 | NP_666948.1 | 1 | 317 | ||
GenBank mRNA | BC127987.1 | - | 321 | 40135591 |