siRNA Details
| Gene Symbol: Olfr1104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146767.2 | NP_666978.2 | 1 | 706 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1104| Gene Symbol: Olfr1104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146767.2 | NP_666978.2 | 1 | 706 | ||