siRNA Details
| Gene Symbol: Gm10081 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162940.1 | NP_001156412.1 | 1 | 516 | ||
| GenBank mRNA | BC139756.1 | - | 1 | 507 | ||
| Gene Symbol: Olfr715 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146780.2 | NP_666991.1 | 1 | 607 | ||
| GenBank mRNA | BC139445.1 | - | 1 | 608 | 8862467 | |
| BC139446.1 | - | 1 | 608 | 8862468 | ||

Gm10081
Genomic Map