siRNA Details
| Gene Symbol: Olfr421-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146720.3 | NP_666931.3 | 1 | 927 | ||
| GenBank mRNA | BC119233.1 | - | 1 | 927 | 8733982 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr421-ps1| Gene Symbol: Olfr421-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146720.3 | NP_666931.3 | 1 | 927 | ||
| GenBank mRNA | BC119233.1 | - | 1 | 927 | 8733982 | |