siRNA Details
| Gene Symbol: Olfr424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104096.1 | - | 1 | 890 | 40043031 | |
| BC104097.1 | - | 1 | 890 | 40043033 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr424| Gene Symbol: Olfr424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC104096.1 | - | 1 | 890 | 40043031 | |
| BC104097.1 | - | 1 | 890 | 40043033 | ||