siRNA Details
Gene Symbol: Olfr1106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146752.2 | NP_666963.2 | 1 | 824 | ||
GenBank mRNA | BC127974.1 | - | 1 | 828 | 40135141 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146752.2 | NP_666963.2 | 1 | 824 | ||
GenBank mRNA | BC127974.1 | - | 1 | 828 | 40135141 |