siRNA Details
| Gene Symbol: Olfr1406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146763.2 | NP_666974.2 | 1 | 887 | ||
| GenBank mRNA | BC139449.1 | - | 1 | 887 | 8862471 | |
| BC139452.1 | - | 1 | 887 | 8862474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1406| Gene Symbol: Olfr1406 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146763.2 | NP_666974.2 | 1 | 887 | ||
| GenBank mRNA | BC139449.1 | - | 1 | 887 | 8862471 | |
| BC139452.1 | - | 1 | 887 | 8862474 | ||