siRNA Details
| Gene Symbol: Olfr1240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146808.2 | NP_667019.1 | 1 | 401 | ||
| GenBank mRNA | BC137782.1 | - | 1 | 401 | 8860801 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1240| Gene Symbol: Olfr1240 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146808.2 | NP_667019.1 | 1 | 401 | ||
| GenBank mRNA | BC137782.1 | - | 1 | 401 | 8860801 | |