siRNA Details
| Gene Symbol: Olfr629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146821.2 | NP_667032.2 | 1 | 586 | ||
| GenBank mRNA | BC120634.1 | - | 1 | 586 | 40129557 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr629| Gene Symbol: Olfr629 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146821.2 | NP_667032.2 | 1 | 586 | ||
| GenBank mRNA | BC120634.1 | - | 1 | 586 | 40129557 | |