siRNA Details
Gene Symbol: Olfr640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146822.2 | NP_667033.2 | 1 | 135 | ||
GenBank mRNA | BC106802.2 | - | 1 | 135 | 40042505 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146822.2 | NP_667033.2 | 1 | 135 | ||
GenBank mRNA | BC106802.2 | - | 1 | 135 | 40042505 |