siRNA Details
| Gene Symbol: Olfr1184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146823.1 | NP_667034.1 | 1 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1184| Gene Symbol: Olfr1184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146823.1 | NP_667034.1 | 1 | 74 | ||