siRNA Details
| Gene Symbol: Olfr273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146824.1 | NP_667035.1 | 1 | 548 | ||
| GenBank mRNA | BC125463.1 | - | 1 | 609 | 40129978 | |
| BC125465.1 | - | 1 | 609 | 40129980 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr273| Gene Symbol: Olfr273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146824.1 | NP_667035.1 | 1 | 548 | ||
| GenBank mRNA | BC125463.1 | - | 1 | 609 | 40129978 | |
| BC125465.1 | - | 1 | 609 | 40129980 | ||