siRNA Details
Gene Symbol: Olfr1205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146896.3 | NP_667107.2 | 2 | 478 | ||
GenBank mRNA | CB173413.1 | - | 1 | 168 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146896.3 | NP_667107.2 | 2 | 478 | ||
GenBank mRNA | CB173413.1 | - | 1 | 168 |