siRNA Details
| Gene Symbol: Olfr1248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146791.2 | NP_667002.2 | 1 | 965 | ||
| GenBank mRNA | BC150718.1 | - | 1 | 967 | 9087629 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1248| Gene Symbol: Olfr1248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146791.2 | NP_667002.2 | 1 | 965 | ||
| GenBank mRNA | BC150718.1 | - | 1 | 967 | 9087629 | |