siRNA Details
Gene Symbol: Olfr1294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146885.1 | NP_667096.1 | 1 | 268 | ||
GenBank mRNA | BC127976.1 | - | 1 | 272 | 40135176 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1294 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146885.1 | NP_667096.1 | 1 | 268 | ||
GenBank mRNA | BC127976.1 | - | 1 | 272 | 40135176 |