siRNA Details
Gene Symbol: Olfr898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146871.2 | NP_667082.2 | 1 | 852 | ||
GenBank mRNA | BC127995.1 | - | 1 | 856 | 40135758 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr898 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146871.2 | NP_667082.2 | 1 | 852 | ||
GenBank mRNA | BC127995.1 | - | 1 | 856 | 40135758 |