siRNA Details
| Gene Symbol: Olfr30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146878.2 | NP_667089.1 | 1 | 929 | ||
| GenBank mRNA | BC119375.1 | - | 1 | 929 | 8734124 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr30| Gene Symbol: Olfr30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146878.2 | NP_667089.1 | 1 | 929 | ||
| GenBank mRNA | BC119375.1 | - | 1 | 929 | 8734124 | |