siRNA Details
| Gene Symbol: Olfr1404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146881.2 | NP_667092.1 | 1 | 867 | ||
| GenBank mRNA | BC120856.1 | - | 1 | 867 | 40129779 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1404| Gene Symbol: Olfr1404 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146881.2 | NP_667092.1 | 1 | 867 | ||
| GenBank mRNA | BC120856.1 | - | 1 | 867 | 40129779 | |