siRNA Details
| Gene Symbol: RPS4Y1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008.3 | NP_000999.1 | 3 | 276 | ||
| XM_017030061.1 | XP_016885550.1 | 3 | 360 | |||
| GenBank mRNA | AK222594.1 | - | 3 | 257 | ||
| AK292819.1 | - | 3 | 261 | |||
| BC010286.1 | - | 2 | 245 | 3048929 | ||
| BC100905.2 | - | 2 | 233 | 40003602 | ||
| BC100906.1 | - | 2 | 231 | |||
| BF242790.1 | - | 3 | 274 | |||
| M58459.1 | - | 2 | 245 | |||

RPS4Y1
Genomic Map