siRNA Details
| Gene Symbol: Olfr1348 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146913.1 | NP_667124.1 | 1 | 438 | ||
| GenBank mRNA | BC120570.1 | - | 1 | 534 | 40129493 | |
| BC137836.1 | - | 1 | 534 | 8860855 | ||
More Information
Set Membership: |
|

Olfr1348
Genomic Map