siRNA Details
Gene Symbol: Olfr1346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146916.1 | NP_667127.1 | 1 | 92 | ||
GenBank mRNA | BC137876.1 | - | 1 | 116 | 8860895 | |
BC137877.1 | - | 1 | 116 | 8860896 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
