siRNA Details
| Gene Symbol: Olfr351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146942.1 | NP_667153.1 | 1 | 265 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr351| Gene Symbol: Olfr351 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146942.1 | NP_667153.1 | 1 | 265 | ||