siRNA Details
| Gene Symbol: RPS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017.2 | NP_001008.1 | 3 | 142 | ||
| GenBank mRNA | AK312060.1 | - | 3 | 136 | ||
| BC000475.1 | - | 1 | 121 | 2961637 | ||
| BC006772.1 | - | 1 | 123 | 2899987 | ||
| BC029732.1 | - | 3 | 122 | 4388273 | ||
| BC066322.1 | - | 3 | 136 | 4816284 | ||
| BC100032.1 | - | 3 | 122 | 6025884 | ||
| L01124.1 | - | 3 | 142 | |||
| L05090.1 | - | 2 | 78 | |||
| X79239.1 | - | 3 | 110 | |||

RPS13
Genomic Map