siRNA Details
Gene Symbol: Olfr1270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146985.2 | NP_667196.1 | 1 | 511 | ||
GenBank mRNA | BC119113.1 | - | 1 | 511 | 8733862 | |
BC119115.1 | - | 1 | 511 | 8733864 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1270 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146985.2 | NP_667196.1 | 1 | 511 | ||
GenBank mRNA | BC119113.1 | - | 1 | 511 | 8733862 | |
BC119115.1 | - | 1 | 511 | 8733864 |