siRNA Details
Gene Symbol: Olfr195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146998.1 | NP_667209.1 | 1 | 462 | ||
GenBank mRNA | BC119110.1 | - | 1 | 485 | 8733859 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr195 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146998.1 | NP_667209.1 | 1 | 462 | ||
GenBank mRNA | BC119110.1 | - | 1 | 485 | 8733859 |