siRNA Details
| Gene Symbol: Olfr172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147001.2 | NP_667212.2 | 1 | 491 | ||
| GenBank mRNA | BC131922.1 | - | 1 | 491 | 40130199 | |
| BC131948.1 | - | 1 | 490 | 40130225 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr172| Gene Symbol: Olfr172 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147001.2 | NP_667212.2 | 1 | 491 | ||
| GenBank mRNA | BC131922.1 | - | 1 | 491 | 40130199 | |
| BC131948.1 | - | 1 | 490 | 40130225 | ||