siRNA Details
| Gene Symbol: Olfr175-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147002.2 | NP_667213.2 | 2 | 806 | ||
| Gene Symbol: Olfr177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146996.2 | NP_667207.2 | 1 | 704 | ||

Olfr175-ps1
Genomic Map