siRNA Details
| Gene Symbol: Olfr392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147006.2 | NP_667217.2 | 2 | 846 | ||
| GenBank mRNA | CB173273.1 | - | 1 | 294 | ||
| CB173600.1 | - | 1 | 537 | |||
| Gene Symbol: Olfr389 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147009.3 | NP_667220.2 | 3 | 1172 | ||

Olfr392
Genomic Map