siRNA Details
| Gene Symbol: Olfr1038-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147013.2 | NP_667224.1 | 2 | 993 | ||
| GenBank mRNA | CB173938.1 | - | 1 | 305 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1038-ps| Gene Symbol: Olfr1038-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147013.2 | NP_667224.1 | 2 | 993 | ||
| GenBank mRNA | CB173938.1 | - | 1 | 305 | ||