siRNA Details
| Gene Symbol: Olfr1234 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146973.2 | NP_667184.2 | 1 | 335 | ||
| GenBank mRNA | BC104287.1 | - | 1 | 335 | ||
| BC104288.1 | - | 1 | 335 | |||
| BC106983.1 | - | 1 | 335 | |||
| BC127132.1 | - | 1 | 335 | 40042541 | ||
| BC127133.1 | - | 1 | 335 | 40042542 | ||

Olfr1234
Genomic Map