siRNA Details
Gene Symbol: Olfr392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147006.2 | NP_667217.2 | 2 | 210 | ||
GenBank mRNA | DR065907.1 | - | 2 | 213 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147006.2 | NP_667217.2 | 2 | 210 | ||
GenBank mRNA | DR065907.1 | - | 2 | 213 |