siRNA Details
| Gene Symbol: Olfr267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146920.2 | NP_667131.2 | 1 | 701 | ||
| GenBank mRNA | BC127983.1 | - | 1 | 704 | 40135361 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr267| Gene Symbol: Olfr267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146920.2 | NP_667131.2 | 1 | 701 | ||
| GenBank mRNA | BC127983.1 | - | 1 | 704 | 40135361 | |