siRNA Details
| Gene Symbol: Olfr631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271020.1 | NP_001257949.1 | 4 | 1365 | ||
| NM_146959.2 | NP_667170.2 | 5 | 1304 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr631| Gene Symbol: Olfr631 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271020.1 | NP_001257949.1 | 4 | 1365 | ||
| NM_146959.2 | NP_667170.2 | 5 | 1304 | |||