siRNA Details
Gene Symbol: Olfr1233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146972.1 | NP_667183.1 | 1 | 869 | ||
GenBank mRNA | BC119149.1 | - | 1 | 917 | 8733898 | |
BC119151.1 | - | 1 | 917 | 8733900 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146972.1 | NP_667183.1 | 1 | 869 | ||
GenBank mRNA | BC119149.1 | - | 1 | 917 | 8733898 | |
BC119151.1 | - | 1 | 917 | 8733900 |