siRNA Details
| Gene Symbol: Olfr447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146988.1 | NP_667199.1 | 1 | 913 | ||
| GenBank mRNA | BC104271.1 | - | 1 | 913 | 40040950 | |
| BC104272.1 | - | 1 | 911 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr447| Gene Symbol: Olfr447 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146988.1 | NP_667199.1 | 1 | 913 | ||
| GenBank mRNA | BC104271.1 | - | 1 | 913 | 40040950 | |
| BC104272.1 | - | 1 | 911 | |||