siRNA Details
Gene Symbol: Olfr1120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147029.1 | NP_667240.1 | 1 | 297 | ||
GenBank mRNA | BC119117.1 | - | 1 | 317 | 8733866 | |
BC119143.1 | - | 1 | 317 | 8733892 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147029.1 | NP_667240.1 | 1 | 297 | ||
GenBank mRNA | BC119117.1 | - | 1 | 317 | 8733866 | |
BC119143.1 | - | 1 | 317 | 8733892 |