siRNA Details
| Gene Symbol: Olfr1122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147031.1 | NP_667242.1 | 1 | 283 | ||
| GenBank mRNA | BC119331.1 | - | 1 | 334 | 8734080 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1122| Gene Symbol: Olfr1122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147031.1 | NP_667242.1 | 1 | 283 | ||
| GenBank mRNA | BC119331.1 | - | 1 | 334 | 8734080 | |