siRNA Details
| Gene Symbol: Olfr600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147046.2 | NP_667257.2 | 1 | 406 | ||
| GenBank mRNA | BC130247.1 | - | 1 | 410 | 40142365 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr600| Gene Symbol: Olfr600 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147046.2 | NP_667257.2 | 1 | 406 | ||
| GenBank mRNA | BC130247.1 | - | 1 | 410 | 40142365 | |