siRNA Details
Gene Symbol: Olfr582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147053.1 | NP_667264.1 | 1 | 673 | ||
GenBank mRNA | BC107338.1 | - | 1 | 673 | ||
BC107339.1 | - | 1 | 673 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147053.1 | NP_667264.1 | 1 | 673 | ||
GenBank mRNA | BC107338.1 | - | 1 | 673 | ||
BC107339.1 | - | 1 | 673 |