siRNA Details
Gene Symbol: Olfr584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147054.1 | NP_667265.1 | 1 | 945 | ||
GenBank mRNA | BC128007.1 | - | 1 | 949 | 40135987 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147054.1 | NP_667265.1 | 1 | 945 | ||
GenBank mRNA | BC128007.1 | - | 1 | 949 | 40135987 |