siRNA Details
Gene Symbol: Olfr641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147072.1 | NP_667283.1 | 1 | 345 | ||
GenBank mRNA | BC127981.1 | - | 1 | 349 | 40135228 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147072.1 | NP_667283.1 | 1 | 345 | ||
GenBank mRNA | BC127981.1 | - | 1 | 349 | 40135228 |