siRNA Details
Gene Symbol: Olfr656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147075.1 | NP_667286.1 | 1 | 93 | ||
GenBank mRNA | BC110472.1 | - | 1 | 93 | 40044290 | |
BC110473.2 | - | 1 | 92 | 40044291 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr656 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147075.1 | NP_667286.1 | 1 | 93 | ||
GenBank mRNA | BC110472.1 | - | 1 | 93 | 40044290 | |
BC110473.2 | - | 1 | 92 | 40044291 |