siRNA Details
| Gene Symbol: Olfr643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147077.1 | NP_667288.1 | 1 | 524 | ||
| GenBank mRNA | BC106803.2 | - | 1 | 524 | 40042567 | |
| BC146607.1 | - | 1 | 524 | 40042594 | ||
| BC148182.1 | - | 1 | 524 | 40043246 | ||

Olfr643
Genomic Map