siRNA Details
| Gene Symbol: Olfr568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147091.2 | NP_667302.2 | 1 | 457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr568| Gene Symbol: Olfr568 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_147091.2 | NP_667302.2 | 1 | 457 | ||